2,2'-Dibromoacetophenone ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-1-(2-bromophenyl)ethanone2-Bromophenacyl bromide

B4124-1G B4124-5G
TCB4124-5GEA 498.21 USD
TCB4124-5G TCB4124-1G
2,2'-Dibromoacetophenone ≥97.0% (by GC)
2,2'-Dibromoacetophenone

CAS Number: 49851-55-0
MDL Number: MFCD00044660
Molecular Formula: C8H6Br2O
Molecular Weight: 277.94
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Specific Gravity (20/20): 1.89


Other Product Names / Synonyms: 2,2'-Dibromoacetophenone

Formula: C₈H₆Br₂O
MW: 277.9406 g/mol
Boiling Pt: 100 °C
Density: 1.89 (20 °C)
MDL Number: MFCD00044660
CAS Number: 49851-55-0
https://us.vwr.com/stibo/web/std.lang.all/54/52/28935452.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR