Benserazide hydrochloride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: DL-Serine 2-(2,3,4-Trihydroxybenzyl)hydrazide Hydrochloride

B4108-1G B4108-5G
TCB4108-1GEA 73.54 USD
TCB4108-1G TCB4108-5G
Benserazide hydrochloride ≥97.0% (by HPLC, titration analysis)
Benserazide hydrochloride

CAS Number: 14919-77-8
MDL Number: MFCD00078571
Molecular Formula: C10H15N3O5
Molecular Weight: 293.70
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 180
Lambda max.: 272 nm (HCl aq.)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: DL-Serine 2-(2,3,4-Trihydroxybenzyl)hydrazide Hydrochloride

Formula: C₁₀H₁₅N₃O₅
MW: 293.70 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00078571
CAS Number: 14919-77-8
https://us.vwr.com/stibo/web/std.lang.all/54/28/28935428.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR