Borane-5-ethyl-2-methylpyridine complex ≥90.0% (by titrimetric analysis)

Synonyms: Borane - 5-Ethyl-2-picoline Complex5-Ethyl-2-methylpyridine Borane5-Ethyl-2-picoline Borane

B4084-100ML
TCB4084-100MLEA 347.43 USD
TCB4084-100ML
Borane-5-ethyl-2-methylpyridine complex ≥90.0% (by titrimetric analysis)
Borane-5-ethyl-2-methylpyridine complex

CAS Number: 1014979-56-6
MDL Number: MFCD16621137
Molecular Formula: C8H14BN
Molecular Weight: 135.02
Purity/Analysis Method: 90.0% (T)
Form: Clear Liquid
Flash Point (°C): 105
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Borane - 5-Ethyl-2-picoline Complex, 5-Ethyl-2-methylpyridine Borane, 5-Ethyl-2-picoline Borane

Formula: C₈H₁₄BN
MW: 135.02 g/mol
Flash Pt: 105 °C
MDL Number: MFCD16621137
CAS Number: 1014979-56-6
https://us.vwr.com/stibo/web/std.lang.all/54/09/28935409.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR