Bromhexine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine Hydrochloride2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine Hydrochloride

B4054-25G
TCB4054-25GEA 37.56 USD
TCB4054-25G
Bromhexine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Bromhexine hydrochloride

CAS Number: 611-75-6
MDL Number: MFCD00056626
Molecular Formula: C14H20Br2N2
Molecular Weight: 412.59
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 234
Lambda max.: 307 nm (Decane)


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine Hydrochloride, 2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine Hydrochloride

Formula: C₁₄H₂₀Br₂N₂
MW: 412.59 g/mol
Melting Pt: 234 °C
MDL Number: MFCD00056626
CAS Number: 611-75-6
https://us.vwr.com/stibo/web/std.lang.all/53/68/28935368.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR