5-tert-Butyl-L-glutamate hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: L-Glutamic Acid 5-tert-Butyl EsterH-Glu(OtBu)-OH

B4045-5G B4045-1G
TCB4045-1GEA 45.68 USD
TCB4045-1G TCB4045-5G
5-tert-Butyl-L-glutamate hydrate ≥98.0% (by titrimetric analysis)
5-tert-Butyl-L-glutamate hydrate

CAS Number: 2419-56-9
MDL Number: MFCD00038580
Molecular Formula: C9H17NO4
Molecular Weight: 203.24
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 182
Specific rotation [a]20/D: 10 deg (C=1, H2O)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: L-Glutamic Acid 5-tert-Butyl Ester, H-Glu(OtBu)-OH

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Melting Pt: 182 °C
MDL Number: MFCD00038580
CAS Number: 2419-56-9
https://us.vwr.com/stibo/web/std.lang.all/53/61/28935361.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR