6-Bromonicotinonitrile ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-5-cyanopyridine

B4039-1G B4039-5G
TCB4039-1GEA 39.59 USD
TCB4039-1G TCB4039-5G
6-Bromonicotinonitrile ≥97.0% (by GC)
6-Bromonicotinonitrile

CAS Number: 139585-70-9
MDL Number: MFCD00234141
Molecular Formula: C6H3BrN2
Molecular Weight: 183.01
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 145


Other Product Names / Synonyms: 6-Bromonicotinonitrile, 6-Bromo-3-pyridinecarbonitrile

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Melting Pt: 145 °C
MDL Number: MFCD00234141
CAS Number: 139585-70-9
https://us.vwr.com/stibo/web/std.lang.all/53/53/28935353.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR