Boscalid ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2-Chloro-N-(4'-chlorobiphenyl-2-yl)nicotinamide

B4038-1G B4038-100MG
TCB4038-1GEA 433.41 USD
TCB4038-1G TCB4038-100MG
Boscalid ≥98.0% (by HPLC, total nitrogen)
Boscalid

CAS Number: 188425-85-6
MDL Number: MFCD06795150
Molecular Formula: C18H12Cl2N2O
Molecular Weight: 343.21
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 149


Product Categories: Bioscience, Fungicide Ingredients
Other Product Names / Synonyms: 2-Chloro-N-(4'-chlorobiphenyl-2-yl)nicotinamide

Formula: C₁₈H₁₂Cl₂N₂O
MW: 343.21 g/mol
Melting Pt: 149 °C
MDL Number: MFCD06795150
CAS Number: 188425-85-6
https://us.vwr.com/stibo/web/std.lang.all/53/51/28935351.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR