(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: tert-Butyl (S)-5-oxopyrrolidine-2-carboxylatetert-Butyl (S)-2-pyrrolidone-5-carboxylate(S)-tert-Butyl 5-oxopyrrolidine-2-carboxylate

B4026-1G B4026-5G
TCB4026-1GEA 19.29 USD
TCB4026-1G TCB4026-5G
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate ≥98.0% (by GC)
(S)-tert-Butyl-5-oxopyrrolidine-2-carboxylate

CAS Number: 35418-16-7
MDL Number: MFCD06659481
Molecular Formula: C9H15NO3
Molecular Weight: 185.22
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 104
Specific rotation [a]20/D: 9 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Chiral Building Blocks, Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Pyroglutamic Acid tert-Butyl Ester, tert-Butyl (S)-5-Oxopyrrolidine-2-carboxylate, (S)-5-Oxopyrrolidine-2-carboxylic Acid tert-Butyl Ester, H-Pyr-OtBu

Formula: C9H15NO3
MW: 185.22 g/mol
Melting Pt: 104 °C
MDL Number: MFCD06659481
CAS Number: 35418-16-7
https://us.vwr.com/stibo/web/std.lang.all/53/44/28935344.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR