2-Bromo-2',4'-difluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-Difluorophenacyl bromide2-Bromo-1-(2,4-difluorophenyl)ethanone

B4020-25G B4020-5G
TCB4020-5GEA 96.43 USD
TCB4020-5G TCB4020-25G
2-Bromo-2',4'-difluoroacetophenone ≥98.0% (by GC)
2-Bromo-2',4'-difluoroacetophenone

CAS Number: 102429-07-2
MDL Number: MFCD00142822
Molecular Formula: C8H5BrF2O
Molecular Weight: 235.03
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 35
Flash Point (°C): 51


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,4-Difluorophenacyl Bromide

Formula: C₈H₅BrF₂O
MW: 235.03 g/mol
Melting Pt: 35 °C
Flash Pt: 51 °C
MDL Number: MFCD00142822
CAS Number: 102429-07-2
https://us.vwr.com/stibo/web/std.lang.all/53/36/28935336.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR