1,4-Bis(4-bromophenyl)-1,3-butadiyne ≥98.0% (by HPLC)

Synonyms: 4',4''-Dibromodiphenylbutadiyne

B4013-200MG
TCB4013-200MGEA 160.68 USD
TCB4013-200MG
1,4-Bis(4-bromophenyl)-1,3-butadiyne ≥98.0% (by HPLC)
1,4-Bis(4-bromophenyl)-1,3-butadiyne

CAS Number: 959-88-6
MDL Number: MFCD00825678
Molecular Formula: C16H8Br2
Molecular Weight: 360.05
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 265

Formula: C₁₆H₈Br₂
MW: 360.05 g/mol
Melting Pt: 265 °C
MDL Number: MFCD00825678
CAS Number: 959-88-6
https://us.vwr.com/stibo/web/std.lang.all/53/31/28935331.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR