(R)-(+)-2-Bromoisovaleric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-(+)-2-Bromo-3-methylbutyric acid(R)-2-Bromoisovaleric acid(R)-2-Bromo-3-methylbutyric acid

B4005-5G
TCB4005-5GEA 198 USD
TCB4005-5G
(R)-(+)-2-Bromoisovaleric acid ≥98.0% (by GC, titration analysis)
(R)-(+)-2-Bromoisovaleric acid

CAS Number: 76792-22-8
MDL Number: MFCD00210113
Molecular Formula: C5H9BrO2
Molecular Weight: 181.03
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 44
Flash Point (°C): 113
Specific rotation [a]20/D: 19 deg (C=2, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-2-Bromoisovaleric Acid

Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 100 °C
Melting Pt: 44 °C
Flash Pt: 113 °C
MDL Number: MFCD00210113
CAS Number: 76792-22-8
https://us.vwr.com/stibo/web/std.lang.all/53/16/28935316.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR