8-Bromoguanosine ≥98.0% (by HPLC, titration analysis)

Synonyms: 2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

B4002-5G B4002-1G
TCB4002-5GEA 122.4 USD
TCB4002-5G TCB4002-1G
8-Bromoguanosine ≥98.0% (by HPLC, titration analysis)
8-Bromoguanosine

CAS Number: 4016-63-1
MDL Number: MFCD00037985
Molecular Formula: C10H12BrN5O5
Molecular Weight: 362.14
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 203
Specific rotation [a]20/D: -28 deg (C=2, DMSO)
Storage Temperature: 0-10°C

Formula: C₁₀H₁₂BrN₅O₅
MW: 362.14 g/mol
Melting Pt: 203 °C
MDL Number: MFCD00037985
CAS Number: 4016-63-1
https://us.vwr.com/stibo/web/std.lang.all/53/11/28935311.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR