5-Benzyl-D-glutamate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
D-Glutamic Acid 5-Benzyl Ester
CAS Number: 2578-33-8
MDL Number: MFCD00063111
Molecular Formula: C12H15NO4
Molecular Weight: 237.26
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 158
Specific rotation [a]20/D: -19 deg (C=1, AcOH)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: D-Glutamic Acid 5-Benzyl Ester, H-D-Glu(OBzl)-OH
Formula:
C₁₂H₁₅NO₄ MW: 237.26 g/mol Melting Pt: 158 °C |
MDL Number:
MFCD00063111 CAS Number: 2578-33-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...