2,5-Bis(trifluoromethyl)benzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,5-Bis(trifluoromethyl)benzene-1-carbonyl chloride

B3980-5G B3980-1G
TCB3980-5GEA 145.15 USD
TCB3980-5G TCB3980-1G
2,5-Bis(trifluoromethyl)benzoyl chloride ≥98.0% (by GC, titration analysis)
2,5-Bis(trifluoromethyl)benzoyl chloride

CAS Number: 393-82-8
MDL Number: MFCD00074997
Molecular Formula: C9H3ClF6O
Molecular Weight: 276.56
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 72
Flash Point (°C): 74

Formula: C₉H₃ClF₆O
MW: 276.57 g/mol
Boiling Pt: 72 °C
Flash Pt: 74 °C
MDL Number: MFCD00074997
CAS Number: 393-82-8
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/52/78/28935278.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR