Bleomycin sulfate (mixture) ≥85% (by HPLC)

Supplier: TCI America

B3972-50MG B3972-10MG
TCB3972-10MGEA 237.8 USD
TCB3972-10MG TCB3972-50MG
Bleomycin sulfate (mixture) ≥85% (by HPLC)
Bleomycin sulfate (mixture)

This is used for pneumonitis model studies.


  • White crystals

Formula: C₅₅H₈₄N₁₇O₂₁S₃·xH₂SO₄
MW: 1512.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070310
CAS Number: 9041-93-4
https://us.vwr.com/stibo/web/std.lang.all/52/70/28935270.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR