Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Supplier: TCI America

Synonyms: Bis(cycloocta-1,5-dien)rhodium(I)-tetrafluoroborat

B3961-100MG B3961-1G
TCB3961-100MGEA 115 USD
TCB3961-100MG TCB3961-1G
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)rhodium (I) tetrafluoroborate

CAS Number: 35138-22-8
MDL Number: MFCD00075045
Molecular Formula: C16H24BF4Rh
Molecular Weight: 406.08
Form: Crystal
Color: Yellow Red
Melting point (°C): 184

Formula: C₁₆H₂₄BF₄Rh
MW: 406.072 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00075045
CAS Number: 35138-22-8
https://us.vwr.com/stibo/web/std.lang.all/52/60/28935260.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR