4'-Benzyloxy-2'-hydroxyacetophenone ≥98.0% (by GC)

Synonyms: 1-(4-(Benzyloxy)-2-hydroxyphenyl)ethanone

B3950-1G
TCB3950-1GEA 156.56 USD
TCB3950-1G
4'-Benzyloxy-2'-hydroxyacetophenone ≥98.0% (by GC)
4'-Benzyloxy-2'-hydroxyacetophenone

CAS Number: 29682-12-0
MDL Number: MFCD00054612
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 105

Formula: C₁₅H₁₄O₃
MW: 242.2699 g/mol
Melting Pt: 105 °C
MDL Number: MFCD00054612
CAS Number: 29682-12-0
https://us.vwr.com/stibo/web/std.lang.all/52/53/28935253.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR