11H-Benzo[b]fluoren-11-one ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 11H-Benzo(b)fluoren-11-one

B3932-5G
TCB3932-5GEA 239.43 USD
TCB3932-5G
11H-Benzo[b]fluoren-11-one ≥98.0% (by GC)
11H-Benzo[b]fluoren-11-one

CAS Number: 3074-03-1
MDL Number: MFCD00215960
Molecular Formula: C17H10O
Molecular Weight: 230.27
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 155

Formula: C₁₇H₁₀O
MW: 230.27 g/mol
Melting Pt: 155 °C
MDL Number: MFCD00215960
CAS Number: 3074-03-1
https://us.vwr.com/stibo/web/std.lang.all/52/40/28935240.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR