2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Benzyl-2-(dimethylamino)-1-[4-(morpholino)phenyl]-1-butanone

B3914-25G B3914-100G
TCB3914-100GEA 177.27 USD
TCB3914-100G TCB3914-25G
2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone ≥98.0% (by HPLC, titration analysis)
2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone

CAS Number: 119313-12-1
MDL Number: MFCD00191775
Molecular Formula: C23H30N2O2
Molecular Weight: 366.51
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 116
Flash Point (°C): 113


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 2-Benzyl-2-(dimethylamino)-1-[4-(morpholino)phenyl]-1-butanone

Formula: C₂₃H₃₀N₂O₂
MW: 366.51 g/mol
Melting Pt: 116 °C
Flash Pt: 113 °C
MDL Number: MFCD00191775
CAS Number: 119313-12-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR