Budesonide ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(+)-16alpha,17alpha-Butylidenedioxy-11beta,21-dihydroxy-1,4-pregnadiene-3,20-dione
CAS Number: 51333-22-3
MDL Number: MFCD00083259
Molecular Formula: C25H34O6
Molecular Weight: 430.54
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: 105 deg (C=0.28, CH2Cl2)
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients, Pregnanes (Glucocorticoids)
Other Product Names / Synonyms: (+)-16alpha,17alpha-Butylidenedioxy-11beta,21-dihydroxy-1,4-pregnadiene-3,20-dione
Formula:
C₂₅H₃₄O₆ MW: 430.54 g/mol |
MDL Number:
MFCD00083259 CAS Number: 51333-22-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...