4-Butylbiphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Butyl-1,1'-Biphenyl

B3888-25G B3888-5G
TCB3888-25GEA 113.63 USD
TCB3888-25G TCB3888-5G
4-Butylbiphenyl ≥98.0% (by GC)
4-Butylbiphenyl

CAS Number: 37909-95-8
MDL Number: MFCD00100665
Molecular Formula: C16H18
Molecular Weight: 210.32
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 129
Melting point (°C): 16
Flash Point (°C): 110
Specific Gravity (20/20): 0.98

Formula: C₁₆H₁₈
MW: 210.32 g/mol
Boiling Pt: 129 °C
Melting Pt: 16 °C
Density: 0.98 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00100665
CAS Number: 37909-95-8
https://us.vwr.com/stibo/web/std.lang.all/52/07/28935207.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR