2-Bromo-5-hydroxypyrazine ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 5-Bromopyrazin-2-ol

B3881-1G B3881-200MG
TCB3881-200MGEA 175 USD
TCB3881-200MG TCB3881-1G
2-Bromo-5-hydroxypyrazine ≥95.0% (by GC)
2-Bromo-5-hydroxypyrazine

CAS Number: 374063-92-0
MDL Number: MFCD06245330
Molecular Formula: C4H3BrN2O
Molecular Weight: 174.99
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 115


Other Product Names / Synonyms: 5-Bromo-2(1H)-pyrazinone, 5-Bromo-2-pyrazinol

Formula: C₄H₃BrN₂O
MW: 174.9800 g/mol
Melting Pt: 115 °C
MDL Number: MFCD06245330
CAS Number: 374063-92-0
https://us.vwr.com/stibo/web/std.lang.all/51/97/28935197.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR