3-Bromophenylsulfur pentafluoride ≥95.0% (by GC)

Supplier: TCI America

B3866-1G
TCB3866-1GEA 185 USD
TCB3866-1G
3-Bromophenylsulfur pentafluoride ≥95.0% (by GC)
3-Bromophenylsulphur pentafluoride

CAS Number: 672-30-0
MDL Number: MFCD03425927
Molecular Formula: C6H4BrF5S
Molecular Weight: 283.05
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 88
Specific Gravity (20/20): 1.86

Formula: C₆H₄BrF₅S
MW: 283.06 g/mol
Boiling Pt: 82 °C
Density: 1.86 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD03425927
CAS Number: 672-30-0
https://us.vwr.com/stibo/web/std.lang.all/51/87/28935187.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR