4-(Bromomethyl)benzophenone ≥96.0% (by GC)

Supplier: TCI America

Synonyms: (4-(Bromomethyl)phenyl)(phenyl)methanone

B3840-5G
TCB3840-5GEA 155.1 USD
TCB3840-5G
4-(Bromomethyl)benzophenone ≥96.0% (by GC)
4-(Bromomethyl)benzophenone

CAS Number: 32752-54-8
MDL Number: MFCD00092076
Molecular Formula: C14H11BrO
Molecular Weight: 275.15
Purity/Analysis Method: 96.0% (GC)
Form: Crystal
Melting point (°C): 112

Formula: C₁₄H₁₁BrO
MW: 275.1405 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00092076
CAS Number: 32752-54-8
https://us.vwr.com/stibo/web/std.lang.all/51/64/28935164.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR