Bromotris(dimethylamino)phosphonium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Bromotris(dimethylamino)phosphonium Hexafluorophosphate

B3838-5G B3838-1G
TCB3838-1GEA 23 USD
TCB3838-1G TCB3838-5G
Bromotris(dimethylamino)phosphonium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Bromotris(dimethylamino)phosphonium hexafluorophosphate

CAS Number: 50296-37-2
MDL Number: MFCD00191864
Molecular Formula: C6H18BrF6N3P2
Molecular Weight: 388.07
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis
Other Product Names / Synonyms: BroP

Formula: C₆H₁₈BrF₆N₃P₂
MW: 388.07 g/mol
MDL Number: MFCD00191864
CAS Number: 50296-37-2
https://us.vwr.com/stibo/web/std.lang.all/51/62/28935162.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR