5-Bromoindole-2-carboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
5-Bromo-1H-indole-2-carboxylic acid
B3836-1G
TCB3836-1GEA
87
USD
TCB3836-1G
5-Bromoindole-2-carboxylic acid ≥98.0% (by titrimetric analysis)
5-Bromoindole-2-carboxylic acid
CAS Number: 7254-19-5
MDL Number: MFCD00022705
Molecular Formula: C9H6BrNO2
Molecular Weight: 240.06
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 280
Storage Temperature:
Formula:
C₉H₆BrNO₂ MW: 240.06 g/mol Melting Pt: 280 °C |
MDL Number:
MFCD00022705 CAS Number: 7254-19-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...