2-Benzyloxyaniline ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(Benzyloxy)aniline
B3796-5G
TCB3796-5GEA
119.51
USD
TCB3796-5G
2-Benzyloxyaniline ≥97.0% (by GC, titration analysis)
2-Benzyloxyaniline
CAS Number: 20012-63-9
MDL Number: MFCD03093873
Molecular Formula: C13H13NO
Molecular Weight: 199.25
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 113
Melting point (°C): 38
Formula:
C₁₃H₁₃NO MW: 199.25 g/mol Boiling Pt: 113 °C Melting Pt: 38 °C |
MDL Number:
MFCD03093873 CAS Number: 20012-63-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...