Bis[2-(4-azidosalicylamido)ethyl]disulfide ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
BASED
B3790-10MG
TCB3790-10MGEA
168
USD
TCB3790-10MG
Bis[2-(4-azidosalicylamido)ethyl]disulfide ≥97.0% (by HPLC)
Bis[2-(4-azidosalicylamido)ethyl]disulfide
CAS Number: 199804-21-2
MDL Number: MFCD01861964
Molecular Formula: C18H18N8O4S2
Molecular Weight: 474.51
Purity/Analysis Method:
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Azide Compounds (Click Chemistry), Bioscience, Crosslinkers
Other Product Names / Synonyms: BASED
Formula:
C₁₈H₁₈N₈O₄S₂ MW: 474.51 g/mol |
MDL Number:
MFCD01861964 CAS Number: 199804-21-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...