α,2-Dibromo-5-fluorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromo-2-(bromomethyl)-4-fluorobenzene2-Bromo-5-fluorobenzyl bromide

B3784-25G B3784-5G
TCB3784-25GEA 81 USD
TCB3784-25G TCB3784-5G
α,2-Dibromo-5-fluorotoluene ≥98.0% (by GC)
α,2-Dibromo-5-fluorotoluene

CAS Number: 112399-50-5
MDL Number: MFCD00042187
Molecular Formula: C7H5Br2F
Molecular Weight: 267.92
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 96
Freezing point (°C): 33

Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Boiling Pt: 96 °C
MDL Number: MFCD00042187
CAS Number: 112399-50-5
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/51/19/28935119.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR