2-Picolyl bromide hydrobromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Bromomethyl)pyridine hydrobromide

B3772-5G
TCB3772-5GEA 81.81 USD
TCB3772-5G
2-Picolyl bromide hydrobromide ≥98.0% (by HPLC, titration analysis)
2-Picolyl bromide hydrobromide

CAS Number: 31106-82-8
MDL Number: MFCD01863544
Molecular Formula: C6H6BrN
Molecular Weight: 252.94
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 149


Other Product Names / Synonyms: 2-Picolyl Bromide Hydrobromide

Formula: C₆H₇Br₂N
MW: 252.94 g/mol
Melting Pt: 149 °C
MDL Number: MFCD01863544
CAS Number: 31106-82-8
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/51/10/28935110.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR