5-Bromo-5-nitro-1,3-dioxane ≥98.0% (by GC) for Biochemical Research

Supplier: TCI America

Synonyms: BromonitrodioxaneBronidox L

B3769-5G
TCB3769-5GEA 120.58 USD
TCB3769-5G
5-Bromo-5-nitro-1,3-dioxane ≥98.0% (by GC) for Biochemical Research
5-Bromo-5-nitro-1,3-dioxane

[for Biochemical Research]
CAS Number: 30007-47-7
MDL Number: MFCD00101855
Molecular Formula: C4H6BrNO4
Molecular Weight: 212.00
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 61
Storage Temperature: 0-10°C

Formula: C₄H₆BrNO₄
MW: 212.00 g/mol
Melting Pt: 61 °C
MDL Number: MFCD00101855
CAS Number: 30007-47-7
https://us.vwr.com/stibo/web/std.lang.all/51/06/28935106.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR