1,2-Benzisothiazol-3(2H)-one ≥98.0% (by GC) for biochemical research

Supplier: TCI America

Synonyms: 1,2-Benzisothiazol-3-one

B3767-5G
TCB3767-5GEA 65.98 USD
TCB3767-5G
1,2-Benzisothiazol-3(2H)-one ≥98.0% (by GC) for biochemical research
1,2-Benzisothiazol-3(2H)-one

[for Biochemical Research]
CAS Number: 2634-33-5
MDL Number: MFCD00127753
Molecular Formula: C7H5NOS
Molecular Weight: 151.18
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 158
Storage Temperature: 0-10°C

Formula: C₇H₅NOS
Melting Pt: 158 °C
MDL Number: MFCD00044001
CAS Number: 2634-33-5
https://us.vwr.com/stibo/web/std.lang.all/51/04/28935104.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR