Bis(NHS)PEG5

Synonyms: Di(N-succinimidyl) 4,7,10,13,16-Pentaoxanonadecanedioate4,7,10,13,16-Pentaoxanonadecanedioic Acid Di(N-succinimidyl) Ester

B3728-25MG
TCB3728-25MGEA 31.08 USD
TCB3728-25MG
Bis(NHS)PEG5
Bis(NHS)PEG5

CAS Number: 756526-03-1
Molecular Formula: C22H32N2O13
Molecular Weight: 532.50
Form: Clear Liquid
Color: Colorless
Storage Temperature: 0°C


Other Product Names / Synonyms: Di(N-succinimidyl) 4,7,10,13,16-Pentaoxanonadecanedioate, 4,7,10,13,16-Pentaoxanonadecanedioic Acid Di(N-succinimidyl) Ester

Formula: C₂₂H₃₂N₂O₁₃
MW: 532.50 g/mol
MDL Number: MFCD11041118
CAS Number: 756526-03-1
https://us.vwr.com/stibo/web/std.lang.all/50/71/28935071.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR