2,8-Bis(trifluoromethyl)quinolin-4-ol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline2,8-Bis(trifluoromethyl)-4-quinolinol

B3714-5G B3714-1G
TCB3714-5GEA 111.18 USD
TCB3714-5G TCB3714-1G
2,8-Bis(trifluoromethyl)quinolin-4-ol ≥98.0% (by GC)
2,8-Bis(trifluoromethyl)quinolin-4-ol

CAS Number: 35853-41-9
MDL Number: MFCD00075091
Molecular Formula: C11H5F6NO
Molecular Weight: 281.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 133

Formula: C11H5F6NO
MW: 281.16 g/mol
Melting Pt: 133 °C
MDL Number: MFCD00075091
CAS Number: 35853-41-9
https://us.vwr.com/stibo/web/std.lang.all/50/61/28935061.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR