(2R,3R,5S,6S)-5,6-Bis(hydroxymethyl)-2,3-dimethoxy-2,3-dimethyl-1,4-dioxane ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(2S,3S,5R,6R)-5,6-Dimethoxy-5,6-dimethyl-1,4-dioxane-2,3-dimethanol
CAS Number: 173371-55-6
Molecular Formula: C10H20O6
Molecular Weight: 236.26
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 121
Specific rotation [a]20/D: -174 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2S,3S,5R,6R)-5,6-Dimethoxy-5,6-dimethyl-1,4-dioxane-2,3-dimethanol
Formula:
C₁₀H₂₀O₆ MW: 236.26 g/mol Melting Pt: 121 °C |
CAS Number:
173371-55-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...