2-Benzyloxy-5-fluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(2-(Benzyloxy)-5-fluorophenyl)boronic acid
B3683-5G
B3683-1G
TCB3683-1GEA
37.08
USD
TCB3683-1G
TCB3683-5G
2-Benzyloxy-5-fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Benzyloxy-5-fluorophenylboronic acid
CAS Number: 779331-47-4
MDL Number: MFCD03788403
Molecular Formula: C13H12BFO3
Molecular Weight: 246.04
Form: Crystal
Color: White
Other Product Names / Synonyms: 2-Benzyloxy-5-fluorobenzeneboronic Acid
Formula:
C₁₃H₁₂BFO₃ MW: 246.0420 g/mol |
MDL Number:
MFCD03788403 CAS Number: 779331-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...