2-Bromo-6-chloro-4-fluoroaniline ≥97.0% (by GC)

Supplier: TCI America

B3681-5G
TCB3681-5GEA 88 USD
TCB3681-5G
2-Bromo-6-chloro-4-fluoroaniline ≥97.0% (by GC)
2-Bromo-6-chloro-4-fluoroaniline

CAS Number: 201849-14-1
MDL Number: MFCD00051707
Molecular Formula: C6H4BrClFN
Molecular Weight: 224.46
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 57
Flash Point (°C): 110

Formula: C₆H₄BrClFN
MW: 224.46 g/mol
Melting Pt: 57 °C
Flash Pt: 110 °C
MDL Number: MFCD00051707
CAS Number: 201849-14-1
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/50/01/28935001.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR