N-Fmoc-D-glutamic acid 5-tert-butyl ester ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 5-tert-Butyl EsterFmoc-D-Glu(OtBu)-OH5-tert-Butyl N-Fmoc-D-glutamate

B3669-5G B3669-1G
TCB3669-5GEA 60.87 USD
TCB3669-5G TCB3669-1G
N-Fmoc-D-glutamic acid 5-tert-butyl ester ≥98.0% (by HPLC, titration analysis)
N-Fmoc-D-glutamic acid 5-tert-butyl ester

Form: Crystal
Melting point (°C): 92
Specific rotation [a]20/D: 9 deg (C=1, MeOH)
Storage Temperature: 0-10°C

Formula: C₂₄H₂₇NO₆
MW: 425.48 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00077055
CAS Number: 104091-08-9
https://us.vwr.com/stibo/web/std.lang.all/49/88/28934988.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR