2-Bromo-3-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America

B3666-1G B3666-5G
TCB3666-1GEA 17.24 USD
TCB3666-1G TCB3666-5G
2-Bromo-3-fluoropyridine ≥98.0% (by GC)
2-Bromo-3-fluoropyridine

CAS Number: 40273-45-8
MDL Number: MFCD04114128
Molecular Formula: C5H3BrFN
Molecular Weight: 175.99
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Specific Gravity (20/20): 1.74

Formula: C₅H₃BrFN
Boiling Pt: 62 °C
Density: 1.74 (20 °C)
MDL Number: MFCD04114128
CAS Number: 40273-45-8
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/49/85/28934985.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR