tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
Synonyms:
(4R,6R)-6-Cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetic Acid tert-Butyl Ester
CAS Number: 125971-94-0
MDL Number: MFCD03093958
Molecular Formula: C14H23NO4
Molecular Weight: 269.34
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 69
Specific rotation [a]20/D: -10 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (4R,6R)-6-Cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetic Acid tert-Butyl Ester
Formula:
C₁₄H₂₃NO₄ MW: 269.34 g/mol Melting Pt: 69 °C |
MDL Number:
MFCD03093958 CAS Number: 125971-94-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...