1-(4-Bromophenyl)-1,2,2-triphenylethylene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Bromo-4-(1,2,2-triphenylethenyl)benzene
B3634-1G
B3634-5G
TCB3634-5GEA
422
USD
TCB3634-5G
TCB3634-1G
1-(4-Bromophenyl)-1,2,2-triphenylethylene ≥98.0% (by GC)
1-(4-Bromophenyl)-1,2,2-triphenylethylene
CAS Number: 34699-28-0
Molecular Formula: C26H19Br
Molecular Weight: 411.34
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 159
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 1-Bromo-4-(1,2,2-triphenylethenyl)benzene
Formula:
C₂₆H₁₉Br MW: 411.34 g/mol Melting Pt: 159 °C |
MDL Number:
MFCD20257904 CAS Number: 34699-28-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...