5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt ≥98.0% (by HPLC, titration analysis) for biochemical research

Supplier: TCI America

Synonyms: 5-Bromo-4-chloro-3-indolyl-b-D-glucuronide sodium salt

B3621-10MG B3621-100MG
TCB3621-100MGEA 283 USD
TCB3621-100MG TCB3621-10MG
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt ≥98.0% (by HPLC, titration analysis) for biochemical research
5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide sodium salt

[for Biochemical Research]
CAS Number: 129541-41-9
MDL Number: MFCD00135782
Molecular Formula: C14H13BrClNO7
Molecular Weight: 444.59
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Glucuronidase Substrates
Other Product Names / Synonyms: 5-Bromo-4-chloro-3-indolyl beta-D-Glucopyranosiduronic Acid Sodium Salt, X-Gluc Sodium Salt, X-GlcA Sodium Salt

Formula: C₁₄H₁₃BrClNO₇
MW: 444.59 g/mol
MDL Number: MFCD00135782
CAS Number: 129541-41-9
https://us.vwr.com/stibo/web/std.lang.all/49/48/28934948.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR