BTQBT ≥98.0% (by total nitrogen basis), purified by sublimation

Synonyms: 4,8-Bis(1,3-dithiol-2-ylidene)-4H,8H-benzo[1,2-c:4,5-c']bis[1,2,5]thiadiazoleBis(1,2,5-thiadiazolo)-p-quinobis(1,3-dithiole)

B3612-100MG
TCB3612-100MGEA 378.42 USD
TCB3612-100MG
BTQBT ≥98.0% (by total nitrogen basis), purified by sublimation
BTQBT

CAS Number: 135704-54-0
Molecular Formula: C12H4N4S6
Molecular Weight: 396.55
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 400


Product Categories: Materials Chemistry, Organic Transistor Materials, Highly Purified Reagents
Other Product Names / Synonyms: 4,8-Bis(1,3-dithiol-2-ylidene)-4H,8H-benzo[1,2-c:4,5-c']bis[1,2,5]thiadiazole, Bis(1,2,5-thiadiazolo)-p-quinobis(1,3-dithiole)

Formula: C₁₂H₄N₄S₆
MW: 396.55 g/mol
Melting Pt: 400 °C
CAS Number: 135704-54-0
https://us.vwr.com/stibo/web/std.lang.all/49/39/28934939.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR