2-(Bromomethyl)phenylboronic acid ≥98.0% (by titrimetric analysis) (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (2-(Bromomethyl)phenyl)metaboric acid 2-(Bromomethyl)benzeneboronic acid Dihydroxy(2-(bromomethyl)phenyl)borane

B3600-1G
TCB3600-1GEA 222 USD
TCB3600-1G
2-(Bromomethyl)phenylboronic acid ≥98.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
2-(Bromomethyl)phenylboronic acid

CAS Number: 91983-14-1
MDL Number: MFCD01318941
Molecular Formula: C7H8BBrO2
Molecular Weight: 214.85
Form: Crystal
Color: White
Melting point (°C): 138
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(Bromomethyl)benzeneboronic Acid

Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
Melting Pt: 138 °C
MDL Number: MFCD01318941
CAS Number: 91983-14-1
https://us.vwr.com/stibo/web/std.lang.all/49/27/28934927.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR