(S)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate ≥97.0% (by total nitrogen basis)
Supplier: TCI America
B3593-200MG
B3593-1G
TCB3593-1GEA
521
USD
TCB3593-1G
TCB3593-200MG
(S)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate ≥97.0% (by total nitrogen basis)
(S)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate
Molecular Weight: 431.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 202
Molecular Formula: C19H17BF7N3
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...