Benzoyl trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Trifluoromethanesulfonic Acid Benzoyl EsterBenzoyl Triflate

B3567-5G
TCB3567-5GEA 585.55 USD
TCB3567-5G
Benzoyl trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Benzoyl trifluoromethanesulfonate

CAS Number: 36967-85-8
Molecular Formula: C8H5F3O4S
Molecular Weight: 254.18
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 114
Storage Temperature: 0°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid Benzoyl Ester, Benzoyl Triflate

Formula: C₈H₅F₃O₄S
MW: 254.18 g/mol
Boiling Pt: 94 °C
Flash Pt: 114 °C
CAS Number: 36967-85-8
https://us.vwr.com/stibo/web/std.lang.all/48/98/28934898.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR