(1R,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(1R,2S)-N1-Boc-1,2-cyclohexanediamine
CAS Number: 364385-54-6
MDL Number: MFCD09952106
Molecular Formula: C11H22N2O2
Molecular Weight: 214.31
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: 5.0 deg (C=1, CH2Cl2)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (1R,2S)-N1-Boc-1,2-cyclohexanediamine, (1R,2S)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1R,2S)-2-Amino-1-(Boc-amino)cyclohexane
Formula:
C₁₁H₂₂N₂O₂ MW: 214.31 g/mol |
MDL Number:
MFCD09952106 CAS Number: 364385-54-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...