Benzylhydrazine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3471-5G
TCB3471-5GEA 121.8 USD
TCB3471-5G
Benzylhydrazine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Benzylhydrazine dihydrochloride

CAS Number: 20570-96-1
MDL Number: MFCD00012921
Molecular Formula: C7H10N2
Molecular Weight: 195.09
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 145

Formula: C₇H₁₀N₂
Melting Pt: 145 °C
MDL Number: MFCD00012921
CAS Number: 20570-96-1
https://us.vwr.com/stibo/web/std.lang.all/48/28/28934828.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR