5-Bromo-6-chloro-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) (contains ca. 10% Ethyl Acetate) for biochemical research

Synonyms: Mag-GalMagenta-Gal

B3469-100MG B3469-20MG
TCB3469-100MGEA 60.77 USD
TCB3469-100MG TCB3469-20MG
5-Bromo-6-chloro-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) (contains ca. 10% Ethyl Acetate) for biochemical research
5-Bromo-6-chloro-3-indolyl-beta-D-galactopyranoside

[for Biochemical Research]
CAS Number: 93863-88-8
MDL Number: MFCD00210022
Molecular Formula: C14H15BrClNO6
Molecular Weight: 408.63
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -46 deg (C=1, MeOH)
Storage Temperature: 0°C


Product Categories: Bioscience, Galactose, ƒÀ-Galactosidase Substrates
Other Product Names / Synonyms: Mag-Gal, Magenta-Gal

Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
MDL Number: MFCD00210022
CAS Number: 93863-88-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR