5-Benzylamino-1-pentanol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-(5-Hydroxypentyl)benzylamineN-(5-Hydroxyamyl)benzylamine

B3461-5G
TCB3461-5GEA 98.42 USD
TCB3461-5G
5-Benzylamino-1-pentanol ≥98.0% (by titrimetric analysis)
5-Benzylamino-1-pentanol

CAS Number: 2937-99-7
MDL Number: MFCD09688387
Molecular Formula: C12H19NO
Molecular Weight: 193.29
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 168
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: N-(5-Hydroxyamyl)benzylamine, N-(5-Hydroxypentyl)benzylamine

Formula: C₁₂H₁₉NO
MW: 193.29 g/mol
Boiling Pt: 168 °C
Density: 1.01 (20 °C)
MDL Number: MFCD09688387
CAS Number: 2937-99-7
https://us.vwr.com/stibo/web/std.lang.all/48/22/28934822.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR